Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Hnrnph3
- Entrez Gene ID
-
432467
- Gene Name
- heterogeneous nuclear ribonucleoprotein H3
- Gene Aliases
-
AA693301, AI666703, Hnrph3
- Chromosome Location
-
Chr.10: 63014650 - 63023889 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC049652.1 |
- |
2 - 3 |
263 |
104 |
BU946493.1 |
- |
2 - 3 |
266 |
104 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
DNA replication helicase 2 homolog (yeast) |
Dna2 |
Dna2l, E130315B21Rik |
-43.6kb
|
Anti-sense |
Chr. - 62946980 - 62974188 on Build GRCm38 |
Downstream |
phenazine biosynthesis-like protein domain containing 2 |
Pbld2 |
3110049J23Rik, Mawbp |
4.5kb
|
Anti-sense |
Chr. - 63024049 - 63058813 on Build GRCm38 |
View Full Product Details