Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Rian
- Entrez Gene ID
-
75745
- Gene Name
- RNA imprinted and accumulated in nucleus
- Gene Aliases
-
5530401N18Rik, C130089L09Rik, Meg8
- Chromosome Location
-
Chr.12: 109603945 - 109661716 on Build GRCm38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AV571224.1 |
- |
3 - 4 |
297 |
|
Chr. 12 - 109617222 - 109637887 [+] on Build GRCm38 |
73 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 370 |
Mir370 |
Mirn370, mir-370, mmu-mir-370 |
-6.8kb
|
Sense |
Chr. - 109618258 - 109618336 on Build GRCm38 |
Downstream |
snoRNA AF357425 |
AF357425 |
MBII-48 |
8.9kb
|
Sense |
Chr. - 109636012 - 109636080 on Build GRCm38 |
View Full Product Details