Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BB280704.2 |
- |
1 |
510 |
|
Chr. 14 - 48672094 - 48672763 [-] on Build GRCm38 |
98 |
GenBankmRNA |
AK044621.1 |
- |
|
1648 |
|
Chr. - - [-] on Build |
98 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
orthodenticle homolog 2 opposite strand 1 |
Otx2os1 |
|
|
Anti-sense |
Chr. - 48669288 - 48793348 on Build GRCm38 |
Downstream |
RIKEN cDNA 4933429O19 gene |
4933429O19Rik |
1700031B11Rik |
206.1kb
|
Anti-sense |
Chr. - 48878873 - 48887112 on Build GRCm38 |
View Full Product Details