Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dnajb4
Entrez Gene ID
67035
Gene Name
DnaJ (Hsp40) homolog, subfamily B, member 4
Gene Aliases
1700029A20Rik, 2010306G19Rik, 5730460G06Rik
Chromosome Location
Chr.3: 152183871 - 152210333 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK160761.1 - - 451 82


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY713166.1 - 1 - 2 451 Chr. 3 - 152179412 - 152210279 [-] on Build GRCm38 82


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream GIPC PDZ domain containing family, member 2 Gipc2 2200002N01Rik, AU021850, Semcap2 -13.5kb Sense Chr. - 152093839 - 152165900 on Build GRCm38
Downstream far upstream element (FUSE) binding protein 1 Fubp1 9530027K12Rik, D3Ertd330e, FBP, Fubp, Fubp4 148.0 Anti-sense Chr. - 152210427 - 152236830 on Build GRCm38

View Full Product Details