Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zmynd19
Entrez Gene ID
67187
Gene Name
zinc finger, MYND domain containing 19
Gene Aliases
2700064H14Rik, AA536891, AU014915, mizip
Chromosome Location
Chr.2: 24949802 - 24960200 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK086718.1 - - 1346 109


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BI690866.1 - 1 128 Chr. 2 - 24960682 - 24961562 [+] on Build GRCm38 109
GenBankmRNA AI429695.1 - 1 Chr. 2 - 24960558 - 24960972 [-] on Build GRCm38 109
GenBankmRNA BB754209.1 - 1 287 Chr. 2 - 24960522 - 24960968 [+] on Build GRCm38 109
GenBankmRNA CD807049.1 - 1 614 Chr. 2 - 24960198 - 24960931 [+] on Build GRCm38 108
GenBankmRNA BB115805.1 - 1 181 Chr. 2 - 24960646 - 24960968 [+] on Build GRCm38 109
GenBankmRNA BQ748001.1 - 1 Chr. 2 - 24960396 - 24960965 [+] on Build GRCm38 110
GenBankmRNA BG083052.2 - 1 385 Chr. 2 - 24960426 - 24961011 [+] on Build GRCm38 109
GenBankmRNA BB528573.2 - 1 505 Chr. 2 - 24960305 - 24960967 [+] on Build GRCm38 109


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Downstream diphthamine biosynethesis 7 Dph7 2810443J12Rik, AW060693, Wdr85 1.2kb Sense Chr. - 24962171 - 24973471 on Build GRCm38

View Full Product Details