The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159934.1 | NP_001153406.1 | 3 - 4 | 717 | 70 |
GenBank mRNA | CF174513.1 | - | 2 - 3 | 430 | 70 |