The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163192.1 | NP_001156664.1 | 3 - 4 | 670 | 79 |
GenBank mRNA | BC157908.1 | - | 3 - 4 | 670 | 79 |
BC172069.1 | - | 3 - 4 | 79 |