The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163192.1 | NP_001156664.1 | 5 - 6 | 898 | 68 |
GenBank mRNA | BC157908.1 | - | 5 - 6 | 898 | 68 |
BC172069.1 | - | 5 - 6 | 898 | 68 |