Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krtap8-1
Entrez Gene ID
16703
Gene Name
keratin associated protein 8-1
Gene Aliases
AI326176
Chromosome Location
Chr.16: 89487374 - 89487952 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010675.1 NP_034805.1 1 - 1 452 108
GenBank mRNA AK133727.1 - 1 - 1 453 108
D86423.1 - 1 - 1 384 108

View Full Product Details