Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zufsp
Entrez Gene ID
72580
Gene Name
zinc finger with UFM1-specific peptidase domain
Gene Aliases
2700019D07Rik
Chromosome Location
Chr.10: 33919142 - 33951347 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028287.2 NP_082563.1 5 - 6 1005 85
GenBank mRNA AK046191.1 - 5 - 6 1008 85
AK052071.1 - 5 - 6 989 85
AK077304.1 - 5 - 6 1053 85
AK077730.1 - 5 - 6 1026 85
AK160928.1 - 5 - 6 1004 85
BC089514.1 - 5 - 6 1018 85

View Full Product Details