Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Frem3
Entrez Gene ID
333315
Gene Name
Fras1 related extracellular matrix protein 3
Gene Aliases
Gm795, NV2
Chromosome Location
Chr.8: 80611039 - 80695557 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167898.1 NP_001161370.1 7 - 8 6200 97

View Full Product Details