Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp2c67
Entrez Gene ID
545288
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 67
Gene Aliases
C730004C24Rik
Chromosome Location
Chr.19: 39608844 - 39649083 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024719.2 NP_001019890.1 6 - 7 977 80
GenBank mRNA AK149389.1 - 6 - 7 988 80
BC092260.1 - 6 - 7 977 80
BC106142.1 - 6 - 7 977 80

Gene Symbol
Cyp2c40
Entrez Gene ID
13099
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 40
Gene Aliases
AI265721, AI662255
Chromosome Location
Chr.19: 39767073 - 39812814 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010004.2 NP_034134.2 6 - 7 1057 80
GenBank mRNA AF047727.1 - 6 - 7 971 80
BC139471.1 - 6 - 7 1057 80
BC139472.1 - 6 - 7 1057 80

Gene Symbol
Cyp2c68
Entrez Gene ID
433247
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 68
Gene Aliases
9030012A22Rik
Chromosome Location
Chr.19: 39688836 - 39741101 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039555.2 NP_001034644.2 6 - 7 1025 80
GenBank mRNA AK136413.1 - 5 - 6 655 80
BC019468.1 - 6 - 7 978 80

Gene Symbol
Cyp2c69
Entrez Gene ID
100043108
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 69
Gene Aliases
AI098658
Chromosome Location
Chr.19: 39842660 - 39886769 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104525.1 NP_001097995.1 6 - 7 1027 80
GenBank mRNA BC145630.1 - 7 - 8 1048 80

View Full Product Details