Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Cyp2c67
- Entrez Gene ID
-
545288
- Gene Name
- cytochrome P450, family 2, subfamily c, polypeptide 67
- Gene Aliases
-
C730004C24Rik
- Chromosome Location
-
Chr.19: 39608844 - 39649083 on Build GRCm38
- Gene Symbol
- Cyp2c40
- Entrez Gene ID
-
13099
- Gene Name
- cytochrome P450, family 2, subfamily c, polypeptide 40
- Gene Aliases
-
AI265721, AI662255
- Chromosome Location
-
Chr.19: 39767073 - 39812814 on Build GRCm38
- Gene Symbol
- Cyp2c69
- Entrez Gene ID
-
100043108
- Gene Name
- cytochrome P450, family 2, subfamily c, polypeptide 69
- Gene Aliases
-
AI098658
- Chromosome Location
-
Chr.19: 39842660 - 39886769 on Build GRCm38
View Full Product Details