Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankrd34a
Entrez Gene ID
545554
Gene Name
ankyrin repeat domain 34A
Gene Aliases
AW060394, Ankrd34
Chromosome Location
Chr.3: 96594826 - 96600126 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024851.3 NP_001020022.2 1 - 2 170 93
GenBank mRNA BC052924.1 - 1 - 2 149 93
BY125165.1 - 1 - 2 170 93

View Full Product Details