Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
B3gnt6
Entrez Gene ID
272411
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6 (core 3 synthase)
Gene Aliases
BC039789, Core-3, Core3
Chromosome Location
Chr.7: 98192415 - 98199475 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081167.1 NP_001074636.1 1 - 2 27 99
GenBank mRNA BC039789.1 - 1 - 2 43 99

View Full Product Details