Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krtap4-8
Entrez Gene ID
665992
Gene Name
keratin associated protein 4-8
Gene Aliases
OTTMUSG00000002541
Chromosome Location
Chr.11: 99780014 - 99780643 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085547.2 NP_001079016.1 1 - 1 148 103

Gene Symbol
Gm11595
Entrez Gene ID
100040276
Gene Name
predicted gene 11595
Gene Aliases
OTTMUSG00000002543
Chromosome Location
Chr.11: 99771714 - 99772913 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126322.1 NP_001119794.1 1 - 1 209 295

View Full Product Details