Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp2d11
Entrez Gene ID
545123
Gene Name
cytochrome P450, family 2, subfamily d, polypeptide 11
Gene Aliases
Cyp2d, P450-2D
Chromosome Location
Chr.15: 82389154 - 82394022 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104531.1 NP_001098001.1 7 - 8 1253 110

View Full Product Details