Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Synpo
Entrez Gene ID
104027
Gene Name
synaptopodin
Gene Aliases
9030217H17Rik, 9130229N11, 9330140I15Rik, AW046661
Chromosome Location
Chr.18: 60593981 - 60660174 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109975.1 NP_001103445.1 1 - 2 196 101
GenBank mRNA AK129267.1 - 1 - 2 87 101
AK141564.1 - 1 - 2 198 101
AK158933.1 - 2 - 3 160 101

View Full Product Details