Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Sult2a1
- Entrez Gene ID
-
20859
- Gene Name
- sulfotransferase family 2A, dehydroepiandrosterone (DHEA)-preferring, member 1
- Gene Aliases
-
ST2A1, Std, Sth1, mSTa1
- Chromosome Location
-
Chr.7: 13796246 - 13837410 on Build GRCm38
- Gene Symbol
- Sult2a2
- Entrez Gene ID
-
100043194
- Gene Name
- sulfotransferase family 2A, dehydroepiandrosterone (DHEA)-preferring, member 2
- Gene Aliases
-
C730007P19Rik, ST, ST2A2, Sth2, mSTa2
- Chromosome Location
-
Chr.7: 13733506 - 13779637 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC125365.1 |
- |
- |
|
110 |
BC125367.1 |
- |
- |
|
110 |
L27121.1 |
- |
- |
|
110 |
View Full Product Details