The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207301.2 | NP_997184.1 | 2 - 3 | 322 | 81 |
GenBank mRNA | AK021091.1 | - | 2 - 3 | 321 | 81 |
AK172079.1 | - | 2 - 3 | 335 | 81 | |
BC031769.1 | - | 2 - 3 | 322 | 81 |