Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Speer4e
- Entrez Gene ID
-
624245
- Gene Name
- spermatogenesis associated glutamate (E)-rich protein 4e
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 14933631 - 14938475 on Build GRCm38
- Gene Symbol
- 4930572O03Rik
- Entrez Gene ID
-
381715
- Gene Name
- spermatogenesis associated glutamate (E)-rich protein pseudogene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 15652288 - 15657059 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_073011.1 |
|
2 - 3 |
349 |
95 |
GenBank mRNA |
AK076955.1 |
- |
2 - 3 |
348 |
95 |
View Full Product Details