Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Speer4e
- Entrez Gene ID
-
624245
- Gene Name
- spermatogenesis associated glutamate (E)-rich protein 4e
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 14933631 - 14938475 on Build GRCm38
- Gene Symbol
- Gm17019
- Entrez Gene ID
-
66773
- Gene Name
- predicted gene 17019
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 15028950 - 15033527 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_182957.1 |
NP_892002.1 |
4 - 5 |
692 |
85 |
- Gene Symbol
- Gm9758
- Entrez Gene ID
-
381714
- Gene Name
- predicted gene 9758
- Gene Aliases
-
ENSMUSG00000033219
- Chromosome Location
-
Chr.5: 14910124 - 14915146 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_198666.3 |
NP_941068.1 |
4 - 5 |
683 |
85 |
GenBank mRNA |
BC048454.1 |
- |
4 - 5 |
683 |
85 |
View Full Product Details