The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159751.1 | NP_001153223.1 | 1 - 2 | 177 | 98 |
GenBank mRNA | AK131928.1 | - | 1 - 2 | 468 | 98 |