Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gm16430
Entrez Gene ID
100040894
Gene Name
predicted gene 16430
Gene Aliases
EG666120
Chromosome Location
Chr.X: 54622211 - 54639016 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166601.1 NP_001160073.1 2 - 3 189 88
GenBank mRNA BC108390.1 - 2 - 3 189 88

Gene Symbol
Gm16405
Entrez Gene ID
100040867
Gene Name
predicted gene 16405
Gene Aliases
EG625802
Chromosome Location
Chr.X: 54531208 - 54548013 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166646.1 NP_001160118.1 - 189 88

Gene Symbol
Slxl1
Entrez Gene ID
75140
Gene Name
Slx-like 1
Gene Aliases
4930527E24Rik, OTTMUSG00000017407
Chromosome Location
Chr.X: 55226876 - 55243736 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029181.1 NP_083457.1 2 - 3 88
GenBank mRNA AK015913.1 - 2 - 3 88
BC108389.1 - - 192 88

View Full Product Details