Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gm16405
Entrez Gene ID
100040867
Gene Name
predicted gene 16405
Gene Aliases
EG625802
Chromosome Location
Chr.X: 54531208 - 54548013 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166646.1 NP_001160118.1 - 320 114

Gene Symbol
Gm16430
Entrez Gene ID
100040894
Gene Name
predicted gene 16430
Gene Aliases
EG666120
Chromosome Location
Chr.X: 54622211 - 54639016 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166601.1 NP_001160073.1 3 - 4 320 114
GenBank mRNA BC108390.1 - 3 - 4 320 114

Gene Symbol
Slxl1
Entrez Gene ID
75140
Gene Name
Slx-like 1
Gene Aliases
4930527E24Rik, OTTMUSG00000017407
Chromosome Location
Chr.X: 55226876 - 55243736 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029181.1 NP_083457.1 3 - 4 355 114
GenBank mRNA AK015913.1 - 3 - 4 356 114
BC108389.1 - - 323 114

View Full Product Details