The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168517.1 | NP_001161989.1 | 1 - 2 | 114 | 106 |
GenBank mRNA | AK172151.1 | - | 1 - 2 | 114 | 106 |
BC056621.1 | - | 1 - 2 | 82 | 106 |