The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172472.1 | NP_001165943.1 | 5 - 6 | 818 | 90 |
NM_001172473.1 | NP_001165944.1 | 5 - 6 | 704 | 90 | |
NM_001172475.1 | NP_001165946.1 | 5 - 6 | 743 | 90 | |
NM_011451.3 | NP_035581.1 | 3 - 4 | 475 | 90 | |
NM_025367.6 | NP_079643.2 | 5 - 6 | 701 | 90 | |
GenBank mRNA | AF068748.1 | - | 5 - 6 | 743 | 90 |
AF068749.1 | - | 3 - 4 | 475 | 90 | |
AF415213.1 | - | 3 - 4 | 373 | 90 | |
AK136289.1 | - | 4 - 5 | 489 | 90 | |
AK155332.1 | - | 5 - 6 | 803 | 90 | |
AK160900.1 | - | 5 - 6 | 694 | 90 | |
BC037710.1 | - | 5 - 6 | 616 | 90 |