Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Pira1
- Entrez Gene ID
-
18722
- Gene Name
- paired-Ig-like receptor A1
- Gene Aliases
-
6M21, Ly89, PIR-A1, Pir
- Chromosome Location
-
Chr.7: 3731633 - 3739870 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_011087.1 |
NP_035217.1 |
- |
50 |
83 |
GenBank mRNA |
U96682.1 |
- |
- |
50 |
83 |
- Gene Symbol
- Pira2
- Entrez Gene ID
-
18725
- Gene Name
- paired-Ig-like receptor A2
- Gene Aliases
-
6M23
- Chromosome Location
-
Chr.7: 3836810 - 3845099 on Build GRCm38
- Gene Symbol
- Pira6
- Entrez Gene ID
-
18729
- Gene Name
- paired-Ig-like receptor A6
- Gene Aliases
-
12M1, Pira10, p91B
- Chromosome Location
-
Chr.7: 4274124 - 4284827 on Build GRCm38
- Gene Symbol
- Pirb
- Entrez Gene ID
-
18733
- Gene Name
- paired Ig-like receptor B
- Gene Aliases
-
Gp91, LIR-3, Lilrb3, PIR-B
- Chromosome Location
-
Chr.7: 3712505 - 3720456 on Build GRCm38
View Full Product Details