Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hspb2
Entrez Gene ID
69253
Gene Name
heat shock protein 2
Gene Aliases
27kDa, 2810021G24Rik, HSP27, MKBP
Chromosome Location
Chr.9: 50751072 - 50752354 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024441.3 NP_077761.3 1 - 2 178 86
GenBank mRNA BC064051.1 - 1 - 2 86

View Full Product Details