Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rpf1
Entrez Gene ID
70285
Gene Name
ribosome production factor 1 homolog (S. cerevisiae)
Gene Aliases
2210420E24Rik, 2310066N05Rik, Bxdc5
Chromosome Location
Chr.3: 146506344 - 146521431 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027332.3 NP_081608.1 4 - 5 485 71
NM_027371.3 NP_081647.2 4 - 5 485 71
GenBank mRNA AK008988.1 - 4 - 5 474 71
AK010079.1 - 4 - 5 482 71
AK162073.1 - 4 - 5 488 71
AK171035.1 - 4 - 5 463 71
AK171079.1 - 4 - 5 474 71
BC056165.1 - 4 - 5 436 71
BC089369.1 - 4 - 5 470 71

View Full Product Details