The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027925.3 | NP_082201.2 | 1 - 2 | 187 | 104 |
GenBank mRNA | AK003534.1 | - | 1 - 2 | 165 | 104 |
AK152986.1 | - | 1 - 2 | 136 | 104 | |
BC048840.1 | - | 1 - 2 | 138 | 104 | |
BC055454.1 | - | 1 - 2 | 150 | 104 | |
BY283216.1 | - | 1 - 2 | 188 | 104 | |
CJ166630.1 | - | 1 - 2 | 150 | 104 |