Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spdyb
Entrez Gene ID
74673
Gene Name
speedy homolog B (Xenopus laevis)
Gene Aliases
4930445A17Rik, 4930451F05Rik, Spdy2, Spdye4
Chromosome Location
Chr.5: 143216316 - 143226902 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029048.3 NP_083324.3 5 - 6 824 103
GenBank mRNA AK015441.1 - 5 - 6 825 103

View Full Product Details