Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cops7b
Entrez Gene ID
26895
Gene Name
COP9 (constitutive photomorphogenic) homolog, subunit 7b (Arabidopsis thaliana)
Gene Aliases
D1Wsu66e, E130114M23
Chromosome Location
Chr.1: 86582902 - 86606500 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172974.2 NP_766562.1 3 - 4 386 95
GenBank mRNA AF071317.1 - 3 - 4 364 95
AK081309.1 - 3 - 4 386 95
AK168740.1 - 3 - 4 345 95

View Full Product Details