Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bpi
Entrez Gene ID
329547
Gene Name
bactericidal permeablility increasing protein
Gene Aliases
9230105K17Rik, Bpifd1
Chromosome Location
Chr.2: 158258091 - 158284533 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177850.3 NP_808518.1 9 - 10 1044 84
GenBank mRNA AK033770.1 - 9 - 10 1046 84
AY363993.1 - 9 - 10 1042 84
AY648037.1 - 9 - 10 1044 84
AY853179.1 - 9 - 10 1029 84
BC118049.1 - 9 - 10 1038 84
BC118504.1 - 9 - 10 1038 84

View Full Product Details