Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Defb50
Entrez Gene ID
387334
Gene Name
defensin beta 50
Gene Aliases
AY387658, BD-50, Defb37, Pbd1, mBD-50
Chromosome Location
Chr.8: 21823539 - 21831296 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199067.1 NP_951022.1 1 - 2 133 112
GenBank mRNA AY387658.1 - 1 - 2 133 112
DQ012050.1 - 1 - 2 133 112

View Full Product Details