Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C1ql3
Entrez Gene ID
227580
Gene Name
C1q-like 3
Gene Aliases
1110065A22Rik, AI661623, C1ql, C1qtnf13, CTRP13, K100
Chromosome Location
Chr.2: 13001887 - 13010864 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153155.2 NP_694795.1 1 - 1 350 83
GenBank mRNA AB044560.1 - - 1233 83
AK053854.1 - - 83
BC024634.1 - 1 - 1 350 83
EU399230.1 - 1 - 1 333 83

View Full Product Details