The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194336.2 | NP_919317.2 | 11 - 11 | 3949 | 93 |
GenBank mRNA | AK128993.1 | - | - | 3944 | 93 |
AK129021.1 | - | - | 3220 | 93 | |
BC007143.1 | - | 10 - 10 | 3805 | 93 | |
BX525828.1 | - | 1 - 1 | 102 | 93 |