Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Mir25
- Entrez Gene ID
-
723926
- Gene Name
- microRNA 25
- Gene Aliases
-
Mirn25, mir-25, mmu-mir-25
- Chromosome Location
-
Chr.5: 138165321 - 138165404 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_029787.1 |
|
1 - 1 |
41 |
56 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AA064230.1 |
- |
1 |
367 |
|
Chr. 5 - 138165307 - 138165718 [-] on Build GRCm38 |
56 |
GenBankmRNA |
CB205037.1 |
- |
1 |
154 |
|
Chr. 5 - 138164676 - 138165517 [-] on Build GRCm38 |
56 |
GenBankmRNA |
AV572397.1 |
- |
1 |
146 |
|
Chr. 5 - 138164898 - 138165504 [-] on Build GRCm38 |
56 |
GenBankmRNA |
AW762527.1 |
- |
1 |
135 |
|
Chr. 5 - 138164782 - 138165498 [-] on Build GRCm38 |
56 |
GenBankmRNA |
AI595464.1 |
- |
1 |
353 |
|
Chr. 5 - 138164802 - 138165716 [-] on Build GRCm38 |
56 |
GenBankmRNA |
AW476402.1 |
- |
1 |
134 |
|
Chr. 5 - 138164718 - 138165497 [-] on Build GRCm38 |
56 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
COP9 (constitutive photomorphogenic) homolog, subunit 6 (Arabidopsis thaliana) |
Cops6 |
Sgn3, VIP/MOV34 |
-1.3kb
|
Anti-sense |
Chr. - 138161102 - 138163984 on Build GRCm38 |
Downstream |
microRNA 93 |
Mir93 |
Mirn93, mir-93, mmu-mir-93 |
119.0
|
Sense |
Chr. - 138165523 - 138165610 on Build GRCm38 |
View Full Product Details