Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dusp6
Entrez Gene ID
67603
Gene Name
dual specificity phosphatase 6
Gene Aliases
1300019I03Rik, MKP-3, MKP3, PYST1
Chromosome Location
Chr.10: 99263231 - 99267489 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026268.3 NP_080544.1 3 - 3 1918 76
GenBank mRNA AK005062.1 - 3 - 3 1919 76
AK009131.1 - 3 - 3 1846 76
AK088468.1 - 3 - 3 1920 76
AK088665.1 - 3 - 3 1920 76
AK148192.1 - 3 - 3 1921 76
AK151126.1 - 3 - 3 1915 76
AK152773.1 - 3 - 3 1915 76
AK159146.1 - 3 - 3 1819 76
AK159502.1 - 3 - 3 1919 76
AK159900.1 - 3 - 3 1819 76
BC003869.1 - 3 - 3 1500 76
BC092272.1 - 3 - 3 1134 76

View Full Product Details