Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Fam19a2
- Entrez Gene ID
-
268354
- Gene Name
- family with sequence similarity 19, member A2
- Gene Aliases
-
6330575M02, FAM19A2 TAFA-2, TAFA2, Tafa-2
- Chromosome Location
-
Chr.10: 123264076 - 123741204 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_045514.1 |
|
4 - 5 |
1852 |
83 |
GenBank mRNA |
AK139403.1 |
- |
4 - 5 |
1936 |
83 |
AK140763.1 |
- |
4 - 5 |
1815 |
83 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
predicted gene 4129 |
Gm4129 |
|
-506.1kb
|
Sense |
Chr. - 123196923 - 123199585 on Build GRCm38 |
Downstream |
eukaryotic translation initiation factor 3, subunit I pseudogene |
4930503E24Rik |
|
738.7kb
|
Sense |
Chr. - 124469074 - 124524488 on Build GRCm38 |
View Full Product Details