The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253738.1 | NP_001240667.1 | 2 - 3 | 475 | 104 |
NM_001253740.1 | NP_001240669.1 | 2 - 3 | 475 | 104 | |
NM_001271764.1 | NP_001258693.1 | 2 - 3 | 475 | 104 | |
NM_001293748.1 | NP_001280677.2 | 3 - 4 | 458 | 104 | |
NM_001293749.1 | NP_001280678.1 | 2 - 3 | 475 | 104 | |
NM_022314.3 | NP_071709.3 | 3 - 4 | 458 | 104 | |
GenBank mRNA | AF317223.1 | - | 3 - 4 | 372 | 104 |
AK050665.1 | - | 2 - 3 | 321 | 104 | |
AK088111.1 | - | 2 - 3 | 335 | 104 | |
AK088902.1 | - | 2 - 3 | 333 | 104 | |
AK145238.1 | - | 2 - 3 | 332 | 104 | |
AK145243.1 | - | 2 - 3 | 332 | 104 | |
AK145804.1 | - | 2 - 3 | 332 | 104 | |
AK146349.1 | - | 2 - 3 | 305 | 104 | |
AK146557.1 | - | 2 - 3 | 457 | 104 | |
AK151441.1 | - | 2 - 3 | 357 | 104 | |
AK151500.1 | - | 2 - 3 | 332 | 104 | |
AK151571.1 | - | 2 - 3 | 331 | 104 | |
AK151578.1 | - | 2 - 3 | 332 | 104 | |
AK151595.1 | - | 2 - 3 | 304 | 104 | |
AK151602.1 | - | 2 - 3 | 332 | 104 | |
AK151991.1 | - | 2 - 3 | 331 | 104 | |
AK152260.1 | - | 2 - 3 | 331 | 104 | |
AK152659.1 | - | 2 - 3 | 304 | 104 | |
AK153031.1 | - | 2 - 3 | 304 | 104 | |
AK160011.1 | - | 2 - 3 | 357 | 104 | |
AK164086.1 | - | 2 - 3 | 332 | 104 | |
AK165428.1 | - | 2 - 3 | 331 | 104 | |
AK166836.1 | - | 2 - 3 | 319 | 104 | |
AK167268.1 | - | 2 - 3 | 309 | 104 | |
AK167534.1 | - | 2 - 3 | 337 | 104 | |
AK168843.1 | - | 2 - 3 | 300 | 104 | |
AK169102.1 | - | 2 - 3 | 303 | 104 | |
AK169245.1 | - | 2 - 3 | 308 | 104 | |
AK169821.1 | - | 2 - 3 | 349 | 104 | |
BC029186.1 | - | 3 - 4 | 415 | 104 | |
BC055407.1 | - | - | 3601 | 104 | |
BC092045.1 | - | 2 - 3 | 308 | 104 | |
CX237327.1 | - | 2 - 3 | 297 | 104 | |
X53753.1 | - | 2 - 3 | 343 | 104 |