Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tpm3
Entrez Gene ID
59069
Gene Name
tropomyosin 3, gamma
Gene Aliases
TM30nm, TMnm, Tm5NM, Tpm-5, Tpm5, Trop-5, gamma-TM, hTM30nm, hTMnm
Chromosome Location
Chr.3: 90072651 - 90100902 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001253738.1 NP_001240667.1 4 - 5 670 82
NM_001271764.1 NP_001258693.1 4 - 5 670 82
NM_022314.3 NP_071709.3 5 - 6 653 82
GenBank mRNA AK050665.1 - 4 - 5 516 82
AK088111.1 - 4 - 5 530 82
AK088902.1 - 4 - 5 528 82
AK145238.1 - 4 - 5 527 82
AK145243.1 - 4 - 5 527 82
AK145804.1 - 4 - 5 527 82
AK146349.1 - 4 - 5 500 82
AK146557.1 - 4 - 5 652 82
AK151441.1 - 4 - 5 552 82
AK151500.1 - 4 - 5 527 82
AK151571.1 - 4 - 5 526 82
AK151578.1 - 4 - 5 527 82
AK151595.1 - 4 - 5 499 82
AK151602.1 - 4 - 5 527 82
AK151991.1 - 4 - 5 526 82
AK152260.1 - 4 - 5 526 82
AK152659.1 - 4 - 5 499 82
AK153031.1 - 4 - 5 499 82
AK160011.1 - 4 - 5 552 82
AK165428.1 - 4 - 5 526 82
AK166836.1 - 4 - 5 514 82
AK167268.1 - 4 - 5 504 82
AK167534.1 - 4 - 5 532 82
AK167581.1 - 4 - 5 527 82
AK168843.1 - 4 - 5 495 82
AK169102.1 - 4 - 5 498 82
AK169245.1 - 4 - 5 503 82
AK169821.1 - 4 - 5 544 82
BC029186.1 - 5 - 6 610 82
BC055407.1 - - 3796 82
BC092045.1 - 4 - 5 503 82
X53753.1 - 4 - 5 538 82

View Full Product Details