Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDUFA6
Entrez Gene ID
101114007
Gene Name
NADH:ubiquinone oxidoreductase subunit A6
Gene Aliases
-
Chromosome Location
Chr.3: 233937819 - 233947660 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027968166.1 XP_027823967.1 1 - 2 173 81

View Full Product Details