Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPL46
Entrez Gene ID
101122386
Gene Name
mitochondrial ribosomal protein L46
Gene Aliases
-
Chromosome Location
Chr.18: 16721705 - 16731681 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004017737.3 XP_004017786.2 3 - 4 846 73

View Full Product Details