Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPL27
Entrez Gene ID
101106861
Gene Name
mitochondrial ribosomal protein L27
Gene Aliases
-
Chromosome Location
Chr.11: 27018431 - 27023520 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012769.3 XP_004012818.1 2 - 3 219 69

View Full Product Details