Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLX1A
Entrez Gene ID
101122755
Gene Name
SLX1 homolog A, structure-specific endonuclease subunit
Gene Aliases
-
Chromosome Location
Chr.24: 26663082 - 26666167 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027962056.1 XP_027817857.1 5 - 6 804 129

View Full Product Details