Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LLPH
Entrez Gene ID
101108472
Gene Name
LLP homolog, long-term synaptic facilitation factor
Gene Aliases
-
Chromosome Location
Chr.3: 164900967 - 164906722 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004006491.4 XP_004006540.1 2 - 3 325 87

View Full Product Details