Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZNRD2
Entrez Gene ID
101108437
Gene Name
zinc ribbon domain containing 2
Gene Aliases
SSSCA1
Chromosome Location
Chr.21: 45547004 - 45548887 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012102131.2 XP_011957521.1 1 - 2 320 84
XM_012102132.2 XP_011957522.1 2 - 3 236 84

View Full Product Details