Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MORN3
Entrez Gene ID
101123583
Gene Name
MORN repeat containing 3
Gene Aliases
-
Chromosome Location
Chr.17: 62566385 - 62581502 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004017347.4 XP_004017396.1 4 - 5 1104 54

View Full Product Details