Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPRC5B
Entrez Gene ID
101112533
Gene Name
G protein-coupled receptor class C group 5 member B
Gene Aliases
-
Chromosome Location
Chr.24: 17665034 - 17686397 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027961692.1 XP_027817493.1 2 - 3 1186 77

View Full Product Details